Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUFIP1
Entrez Gene ID
101103075
Gene Name
nuclear FMR1 interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.10: 17365125 - 17397885 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027973546.1 XP_027829347.1 7 - 8 1019 88

View Full Product Details