Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM83F
Entrez Gene ID
101102139
Gene Name
family with sequence similarity 83 member F
Gene Aliases
-
Chromosome Location
Chr.3: 232182687 - 232219156 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012175687.3 XP_012031077.2 2 - 3 655 64

View Full Product Details