Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNN2
Entrez Gene ID
101105660
Gene Name
potassium calcium-activated channel subfamily N member 2
Gene Aliases
-
Chromosome Location
Chr.7: 3550338 - 3928616 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971450.1 XP_027827251.1 7 - 8 2246 103

View Full Product Details