Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB24
Entrez Gene ID
101113501
Gene Name
zinc finger and BTB domain containing 24
Gene Aliases
-
Chromosome Location
Chr.8: 31037096 - 31048058 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011211.3 XP_004011260.2 5 - 6 1384 68

View Full Product Details