Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGK2
Entrez Gene ID
101107121
Gene Name
serum/glucocorticoid regulated kinase 2
Gene Aliases
-
Chromosome Location
Chr.13: 75293388 - 75320158 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004014577.4 XP_004014626.1 4 - 5 538 98
XM_027977208.1 XP_027833009.1 3 - 4 228 98
XM_027977209.1 XP_027833010.1 3 - 4 228 98
XM_027977210.1 XP_027833011.1 3 - 4 228 98
XM_027977211.1 XP_027833012.1 3 - 4 228 98
XM_027977213.1 XP_027833014.1 3 - 4 228 98
XM_027977214.1 XP_027833015.1 3 - 4 391 98

View Full Product Details