Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFDC13
Entrez Gene ID
101112081
Gene Name
WAP four-disulfide core domain 13
Gene Aliases
-
Chromosome Location
Chr.13: 77837057 - 77841656 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976406.1 XP_027832207.1 1 - 2 85 94

View Full Product Details