Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCD
Entrez Gene ID
101113102
Gene Name
IQ motif containing D
Gene Aliases
-
Chromosome Location
Chr.17: 70226179 - 70249015 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017395.4 XP_004017444.3 2 - 3 949 137

View Full Product Details