Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TGM7
Entrez Gene ID
101106629
Gene Name
transglutaminase 7
Gene Aliases
-
Chromosome Location
Chr.18: 55352968 - 55367254 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957271.1 XP_027813072.1 12 - 13 2065 57

View Full Product Details