Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM71
Entrez Gene ID
101112682
Gene Name
tripartite motif containing 71
Gene Aliases
-
Chromosome Location
Chr.19: 7801210 - 7861864 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957910.1 XP_027813711.1 2 - 3 1043 95

View Full Product Details