Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPRC5C
Entrez Gene ID
101105426
Gene Name
G protein-coupled receptor class C group 5 member C
Gene Aliases
-
Chromosome Location
Chr.11: 6272375 - 6289339 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015098972.2 XP_014954458.2 2 - 3 1192 111

View Full Product Details