Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IHH
Entrez Gene ID
101113658
Gene Name
Indian hedgehog signaling molecule
Gene Aliases
-
Chromosome Location
Chr.2: 235819734 - 235825989 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965204.1 XP_027821005.1 2 - 3 734 64

View Full Product Details