Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDYL2
Entrez Gene ID
101119749
Gene Name
chromodomain Y like 2
Gene Aliases
-
Chromosome Location
Chr.14: 7577965 - 7744742 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977652.1 XP_027833453.1 5 - 6 1605 72
XM_027977653.1 XP_027833454.1 5 - 6 1411 72

View Full Product Details