Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNBD1
Entrez Gene ID
101112328
Gene Name
cyclic nucleotide binding domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 97287231 - 97886925 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027973294.1 XP_027829095.1 1 - 2 82 103

View Full Product Details