Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BOLA2B
Entrez Gene ID
101114744
Gene Name
bolA family member 2B
Gene Aliases
-
Chromosome Location
Chr.24: 26666006 - 26667278 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962060.1 XP_027817861.1 2 - 3 741 102

View Full Product Details