Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
MAP3K10
Entrez Gene ID
101118723
Gene Name
mitogen-activated protein kinase kinase kinase 10
Gene Aliases
-
Chromosome Location
Chr.14: 53085588 - 53101944 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978331.1 XP_027834132.1 2 - 3 1269 102

View Full Product Details