Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC1B
Entrez Gene ID
101119293
Gene Name
C-type lectin domain family 1 member B
Gene Aliases
-
Chromosome Location
Chr.3: 220203410 - 220214036 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012175411.3 XP_012030801.2 2 - 3 238 109

View Full Product Details