Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD10
Entrez Gene ID
101120519
Gene Name
BTB domain containing 10
Gene Aliases
-
Chromosome Location
Chr.15: 42245105 - 42322649 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004016140.4 XP_004016189.1 4 - 5 991 110
XM_012095724.2 XP_011951114.1 5 - 6 897 110
XM_012095725.3 XP_011951115.1 6 - 7 973 110
XM_012095726.3 XP_011951116.1 6 - 7 976 110
XM_012095734.2 XP_011951124.1 4 - 5 742 110
XM_027979369.1 XP_027835170.1 5 - 6 1601 110
XM_027979371.1 XP_027835172.1 6 - 7 1043 110
XM_027979372.1 XP_027835173.1 5 - 6 897 110
XM_027979373.1 XP_027835174.1 6 - 7 973 110
XM_027979374.1 XP_027835175.1 4 - 5 770 110

View Full Product Details