Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNURF
Entrez Gene ID
105601857
Gene Name
SNRPN upstream reading frame
Gene Aliases
-
Chromosome Location
Chr.18: 1116622 - 1139527 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012098298.2 XP_011953688.1 2 - 3 206 75

View Full Product Details