Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIPI
Entrez Gene ID
101122756
Gene Name
lipase I
Gene Aliases
-
Chromosome Location
Chr.1: 154510355 - 154590268 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959102.1 XP_027814903.1 12 - 13 1711 131

View Full Product Details