Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TANGO2
Entrez Gene ID
101121122
Gene Name
transport and golgi organization 2 homolog
Gene Aliases
C17H22orf25
Chromosome Location
Chr.17: 82404795 - 82422348 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027956937.1 XP_027812738.1 8 - 9 1183 95
XM_027956939.1 XP_027812740.1 8 - 9 775 95

View Full Product Details