Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA4
Entrez Gene ID
101118993
Gene Name
serpin family A member 4
Gene Aliases
-
Chromosome Location
Chr.18: 59611031 - 59618416 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015102035.2 XP_014957521.2 2 - 3 924 76

View Full Product Details