Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD2
Entrez Gene ID
101115935
Gene Name
FERM and PDZ domain containing 2
Gene Aliases
-
Chromosome Location
Chr.25: 44468380 - 44567251 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015104497.2 XP_014959983.2 12 - 13 1747 100
XM_027962238.1 XP_027818039.1 13 - 14 1834 100
XM_027962240.1 XP_027818041.1 13 - 14 1834 100
XM_027962241.1 XP_027818042.1 13 - 14 1834 100
XM_027962242.1 XP_027818043.1 13 - 14 1834 100

View Full Product Details