Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBP2
Entrez Gene ID
101108710
Gene Name
retinol binding protein 2
Gene Aliases
-
Chromosome Location
Chr.1: 272262368 - 272295063 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004003298.4 XP_004003347.1 3 - 4 419 60

View Full Product Details