Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAL4
Entrez Gene ID
101106580
Gene Name
dynein axonemal light chain 4
Gene Aliases
-
Chromosome Location
Chr.3: 231229315 - 231241146 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006998.4 XP_004007047.1 3 - 4 392 103

View Full Product Details