Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RANBP3L
Entrez Gene ID
101109211
Gene Name
RAN binding protein 3 like
Gene Aliases
-
Chromosome Location
Chr.16: 39961538 - 40019784 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012097051.2 XP_011952441.2 5 - 6 765 93
XM_012097052.2 XP_011952442.2 5 - 6 495 93
XM_027980109.1 XP_027835910.1 5 - 6 558 93
XM_027980110.1 XP_027835911.1 4 - 5 897 93
XM_027980111.1 XP_027835912.1 4 - 5 897 93
XM_027980112.1 XP_027835913.1 4 - 5 897 93
XM_027980113.1 XP_027835914.1 4 - 5 897 93
XM_027980115.1 XP_027835916.1 4 - 5 897 93

View Full Product Details