Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLIPR1L2
Entrez Gene ID
101123622
Gene Name
GLIPR1 like 2
Gene Aliases
-
Chromosome Location
Chr.3: 118539837 - 118575863 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965801.1 XP_027821602.1 2 - 3 476 128

View Full Product Details