Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTSR2
Entrez Gene ID
101104053
Gene Name
neurotensin receptor 2
Gene Aliases
-
Chromosome Location
Chr.3: 21227555 - 21235356 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007210.3 XP_004007259.2 3 - 4 962 95

View Full Product Details