Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAPC5
Entrez Gene ID
101102065
Gene Name
small nuclear RNA activating complex polypeptide 5
Gene Aliases
-
Chromosome Location
Chr.7: 14263062 - 14270537 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010251.4 XP_004010300.1 3 - 4 1079 110
XM_012180801.3 XP_012036191.1 3 - 4 1093 110
XM_027971516.1 XP_027827317.1 2 - 3 1006 110

View Full Product Details