Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPSL
Entrez Gene ID
101110872
Gene Name
calcyphosine like
Gene Aliases
-
Chromosome Location
Chr.16: 40251198 - 40288495 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012097064.3 XP_011952454.1 5 - 6 765 115

View Full Product Details