Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KCTD7
Entrez Gene ID
101110559
Gene Name
potassium channel tetramerization domain containing 7
Gene Aliases
-
Chromosome Location
Chr.24: 28923636 - 28934436 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961768.1 XP_027817569.1 4 - 5 883 73

View Full Product Details