Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FKBP1A
Entrez Gene ID
101105938
Gene Name
FKBP prolyl isomerase 1A
Gene Aliases
-
Chromosome Location
Chr.13: 62064513 - 62090535 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015099799.2 XP_014955285.2 2 - 3 301 68
XM_027977025.1 XP_027832826.1 2 - 3 301 68

View Full Product Details