Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101108364
Entrez Gene ID
101108364
Gene Name
synaptopodin 2-like protein
Gene Aliases
-
Chromosome Location
Chr.25: 30712050 - 30720162 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015104449.2 XP_014959935.2 1 - 2 285 92

View Full Product Details