Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JPT1
Entrez Gene ID
101111302
Gene Name
Jupiter microtubule associated homolog 1
Gene Aliases
HN1
Chromosome Location
Chr.11: 6954559 - 6968767 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013133.4 XP_004013182.1 4 - 5 439 111

View Full Product Details