Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AMZ1
Entrez Gene ID
101111338
Gene Name
archaelysin family metallopeptidase 1
Gene Aliases
-
Chromosome Location
Chr.24: 41447989 - 41455509 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961672.1 XP_027817473.1 5 - 6 1238 67
XM_027961673.1 XP_027817474.1 4 - 5 1068 67

View Full Product Details