Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
APRT
Entrez Gene ID
101108335
Gene Name
adenine phosphoribosyltransferase
Gene Aliases
-
Chromosome Location
Chr.14: 14793588 - 14796220 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015100161.2 XP_014955647.2 1 - 2 167 95

View Full Product Details