Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RDH5
Entrez Gene ID
101115199
Gene Name
retinol dehydrogenase 5
Gene Aliases
-
Chromosome Location
Chr.3: 175432388 - 175440587 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012174796.2 XP_012030186.2 2 - 3 186 66
XM_027967616.1 XP_027823417.1 2 - 3 234 66

Gene Symbol
BLOC1S1
Entrez Gene ID
101105150
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 1
Gene Aliases
-
Chromosome Location
Chr.3: 175436917 - 175440627 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012174798.3 XP_012030188.1 2 - 3 274 66

View Full Product Details