Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF11
Entrez Gene ID
101117833
Gene Name
Kruppel like factor 11
Gene Aliases
-
Chromosome Location
Chr.3: 19894611 - 19906039 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005679.4 XP_004005728.1 2 - 3 436 82
XM_027966514.1 XP_027822315.1 2 - 3 336 82
XM_027966515.1 XP_027822316.1 2 - 3 365 82

View Full Product Details