Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNC4
Entrez Gene ID
101105210
Gene Name
potassium voltage-gated channel subfamily C member 4
Gene Aliases
-
Chromosome Location
Chr.1: 92100518 - 92122799 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972888.1 XP_027828689.1 3 - 4 2244 67

View Full Product Details