Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
GALR3
Entrez Gene ID
101119389
Gene Name
galanin receptor 3
Gene Aliases
-
Chromosome Location
Chr.3: 230466729 - 230472665 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093987.2 XP_014949473.2 1 - 2 4548 132

View Full Product Details