Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPCS3
Entrez Gene ID
101105386
Gene Name
signal peptidase complex subunit 3
Gene Aliases
-
Chromosome Location
Chr.26: 7477917 - 7487659 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015104550.2 XP_014960036.2 2 - 3 1127 105

View Full Product Details