Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DSTN
Entrez Gene ID
101119923
Gene Name
destrin, actin depolymerizing factor
Gene Aliases
-
Chromosome Location
Chr.13: 39320086 - 39346285 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015099645.2 XP_014955131.1 3 - 4 859 87

View Full Product Details