Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC8C
Entrez Gene ID
101121640
Gene Name
leucine rich repeat containing 8 VRAC subunit C
Gene Aliases
-
Chromosome Location
Chr.1: 70844280 - 70927105 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002179.4 XP_004002228.1 2 - 3 343 65

View Full Product Details