Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC9A
Entrez Gene ID
101119033
Gene Name
C-type lectin domain containing 9A
Gene Aliases
-
Chromosome Location
Chr.3: 220186688 - 220197725 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006876.4 XP_004006925.3 5 - 6 697 94
GenBank mRNA KU641038.1 - 5 - 6 595 94

View Full Product Details