Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSSK4
Entrez Gene ID
101115710
Gene Name
testis specific serine kinase 4
Gene Aliases
-
Chromosome Location
Chr.7: 22270643 - 22280313 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012180938.3 XP_012036328.1 1 - 2 986 98

View Full Product Details