Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101118224
Entrez Gene ID
101118224
Gene Name
corticosteroid-binding globulin
Gene Aliases
CBG, SERPINA6
Chromosome Location
Chr.18: 59355950 - 59380111 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017972.4 XP_004018021.3 3 - 4 969 76

View Full Product Details