Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDZD8
Entrez Gene ID
101121648
Gene Name
PDZ domain containing 8
Gene Aliases
-
Chromosome Location
Chr.22: 40765845 - 40844939 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012103001.3 XP_011958391.3 4 - 5 1457 79

View Full Product Details