Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PI16
Entrez Gene ID
101122471
Gene Name
peptidase inhibitor 16
Gene Aliases
-
Chromosome Location
Chr.20: 12503113 - 12517149 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958945.1 XP_027814746.1 5 - 6 1283 95

View Full Product Details