Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMEM86B
Entrez Gene ID
101121701
Gene Name
transmembrane protein 86B
Gene Aliases
-
Chromosome Location
Chr.14: 66916534 - 66918753 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012094771.3 XP_011950161.2 2 - 3 648 74

View Full Product Details