Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FANCB
Entrez Gene ID
101123188
Gene Name
FA complementation group B
Gene Aliases
-
Chromosome Location
Chr.X: 13714606 - 13736778 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963214.1 XP_027819015.1 5 - 6 1511 75

View Full Product Details