Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LLGL1
Entrez Gene ID
101120930
Gene Name
LLGL scribble cell polarity complex component 1
Gene Aliases
-
Chromosome Location
Chr.11: 28899048 - 28913827 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015098677.2 XP_014954163.2 16 - 17 2251 79
XM_027974801.1 XP_027830602.1 16 - 17 2242 79
XM_027974802.1 XP_027830603.1 16 - 17 2239 79
XM_027974803.1 XP_027830604.1 16 - 17 2242 79

View Full Product Details